Novus Biologicals
Manufacturer Code:NBP227163
Catalog # NBP227163
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A portion of amino acid 20-70 of human PU.1 was used as the immunogen. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 31 kDa transforming protein; 31 kDa-transforming protein; hematopoietic transcription factor PU.1; hematopoietic transcription factor PU.1 OF PU.1 SFPI131 kDa transforming protein SPI-1 SPI-1 proto-oncogene SPI-A spleen focus forming virus (SFFV) proviral integration oncogene spi131 kDa-transforming protein transcription factor PU.1; spleen focus forming virus (SFFV) proviral integration oncogene spi1; Transcription factor PU.1
Gene Aliases: OF; PU.1; SFPI1; SPI-1; SPI-A; SPI1
UniProt ID: (Human) P17947
Entrez Gene ID: (Human) 6688
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.