Invitrogen
Manufacturer Code:PA5115807
Catalog # PIPA5115807
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PU.1/Spi1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 31 kDa transforming protein; 31 kDa transforming protein 31 kDa-transforming protein Dis1 Dis-1 hCG_25181 hematopoietic transcription factor PU.1 OF PU.1 SFFV proviral integration 1 SFFV proviral integration 1 protein Sfpi1 Sfpi-1 Spi1 SPI-1 Spi-1 proto-oncogene SPI-A spleen focus forming virus (SFFV) proviral integration oncogene spleen focus forming virus (SFFV) proviral integration oncogene spi1 spleen focus forming virus proviral integration oncogene spi1 Tcfpu1 Tfpu.1 transcription factor PU.1; 31 kDa-transforming protein; hematopoietic transcription factor PU.1; SFFV proviral integration 1; SFFV proviral integration 1 protein; spleen focus forming virus (SFFV) proviral integration oncogene spi1; spleen focus forming virus proviral integration oncogene spi1; Transcription factor PU.1
Gene Aliases: Dis-1; Dis1; OF; PU.1; Sfpi-1; SFPI1; SPI-1; SPI-A; SPI1; Tcfpu1; Tfpu.1
UniProt ID: (Human) P17947, (Mouse) P17433, (Rat) Q6BDS1
Entrez Gene ID: (Human) 6688, (Mouse) 20375, (Rat) 366126
Molecular Function: nucleic acid binding signaling molecule transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.