Invitrogen
Manufacturer Code:PA564410
Catalog # PIPA564410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PTX3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: long pentraxin 3; Pentaxin-related protein PTX3; Pentraxin-related protein PTX3; Pentraxin-related protein PTX3 Pentaxin-related protein PTX3 Tumor necrosis factor alpha-induced protein 5 TNF alpha-induced protein 5 Tumor necrosis factor-inducible gene 14 protein TSG-14 TNFAIP5 TSG14 PTX3; TNF alpha-induced protein 5; TSG-14; Tumor necrosis factor alpha-induced protein 5; Tumor necrosis factor-inducible gene 14 protein; tumor necrosis factor-inducible protein TSG-14
Gene Aliases: PTX3; TNFAIP5; TSG-14; TSG14
UniProt ID: (Human) P26022
Entrez Gene ID: (Human) 5806
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.