Novus Biologicals
Manufacturer Code:NBP29402620UL
Catalog # NB022617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 40-179 of human PTRH2 (NP_057161.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Bcl-2 inhibitor of transcription 1; Bcl-2 inhibitor of transcription 1 BIT1CGI-147 EC 3.1.1.29 mitochondrial peptidyl-tRNA hydrolase 2 PTH2FLJ32471; cilia and flagella associated protein 37; Peptidyl-tRNA hydrolase 2, mitochondrial; PTH 2
Gene Aliases: BIT1; CFAP37; CGI-147; IMNEPD; PTH; PTH 2; PTH2; PTRH2
UniProt ID: (Human) Q9Y3E5
Entrez Gene ID: (Human) 51651
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.