Invitrogen
Manufacturer Code:PA568780
Catalog # PIPA568780
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of mouse PTRF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cav-p60; Caveolae-associated protein 1; cavin; Cavin-1; Polymerase I and transcript release factor; PTRF; PTRF Polymerase I and transcript release factor FKSG13 CAVIN CAVIN1 CGL4 cavin-1 TTF-I interacting peptide 12; ptrf {ECO:0000312|RGD:1307685}; RNA polymerase I and transcript release factor; TTF-I interacting peptide 12
Gene Aliases: 2310075E07Rik; AW546441; Cav-p60; CAVIN; cavin-1; CAVIN1; CGL4; FKSG13; PTRF
UniProt ID: (Human) Q6NZI2, (Mouse) O54724
Entrez Gene ID: (Dog) 100855435, (Bovine) 538457, (Horse) 100066120, (Rabbit) 103351573, (Human) 284119, (Rat) 287710, (Mouse) 19285
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.