Invitrogen
Manufacturer Code:PA127632
Catalog # PIPA127632
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to human Tyrosine phosphatase zeta polypeptide 2 HTPZP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C,Store at 4°C short term. For long term storage,avoiding freeze/thaw cycles. |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: HTPZP2 PTPRZ; protein tyrosine phosphatase, receptor-type, Z polypeptide 1; protein tyrosine phosphatase, receptor-type, zeta polypeptide 1; Protein-tyrosine phosphatase receptor type Z polypeptide 1; Protein-tyrosine phosphatase receptor type Z polypeptide 2; R-PTP-zeta; R-PTP-zeta-2; receptor-type tyrosine phosphatase beta/zeta; Receptor-type tyrosine-protein phosphatase zeta
Gene Aliases: HPTPZ; HPTPzeta; HTPZP2; phosphacan; PTP-ZETA; PTP18; PTPRZ; PTPRZ1; PTPRZ2; PTPZ; R-PTP-zeta-2; RPTPB; RPTPbeta
UniProt ID: (Human) P23471
Entrez Gene ID: (Human) 5803
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.