Proteintech
Manufacturer Code:130081AP150UL
Catalog # 50-172-8415
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PTPRS Fusion Protein Ag3691 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LAR-PTP2; Leukocyte common antigen-related protein-tyrosine phosphatase 2; leukocyte common antigen-related proten-tyrosine phosphatase 2; protein tyrosine phosphatase PTPsigma; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, S; PTPNU-3; PTPRS PTPSIGMA R PTP S R PTP sigma rPTPsigma; R-PTP-S; R-PTP-sigma; Receptor-type tyrosine-protein phosphatase S; Receptor-type tyrosine-protein phosphatase sigma; tissue-specific protein tyrosine phosphatase
Gene Aliases: AL022616; Larptp2; PTP; PTP-NU3; PTPNU-3; Ptprd; PTPRS; PTPSIGMA; Ptpsigma.; Ptpt9; R-PTP-S; R-PTP-sigma; Rptpd; RPTPsigma
UniProt ID: (Human) Q13332, (Mouse) B0V2N1, (Rat) Q64605
Entrez Gene ID: (Human) 5802, (Mouse) 19280, (Rat) 25529
Molecular Function: hydrolase phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.