Invitrogen
Manufacturer Code:PA575158
Catalog # PIPA575158
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PTPRO. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glomerular epithelial protein 1; osteoclastic transmembrane protein-tyrosine phosphatase; phosphotyrosine phosphatase U2; protein tyrosine phosphatase PTP-U2; Protein tyrosine phosphatase U2; protein tyrosine phosphatase, non-receptor type 15; protein tyrosine phosphatase, receptor type, O; PTP phi; PTP-U2; PTPase U2; R-PTP-O; receptor protein tyrosine phosphatase RO; receptor-type protein tyrosine phosphatase D30; Receptor-type tyrosine-protein phosphatase O; Receptor-type tyrosine-protein phosphatase U R-PTP-U Pancreatic carcinoma phosphatase 2 PCP-2 Protein-tyrosine phosphatase J PTP-J hPTP-J Protein-tyrosine phosphatase pi PTP pi Protein-tyrosine phosphatase receptor omicron PTP-RO Receptor-type protein-tyrosine phosphatase psi R-PTP-psi PTPRU FMI PCP2 PTPRO
Gene Aliases: D28; GLEPP1; NPHS6; PTP-OC; PTP-U2; PTPBK; Ptpn15; PTPoc; PTPphi; PTPRO; PTPROT; PTPU2; R-PTP-O; RPTP-BK
UniProt ID: (Human) Q16827, (Mouse) E9Q612
Entrez Gene ID: (Human) 5800, (Rat) 50677, (Mouse) 19277
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.