Novus Biologicals
Manufacturer Code:NBP295232100UL
Catalog # NB024626
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 30-300 of human PTPRO (NP_109592.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.3.48 GLEPP1PTPROt Glomerular epithelial protein 1 phosphotyrosine phosphatase U2 protein tyrosine phosphatase PTP-U2 Protein tyrosine phosphatase U2 protein tyrosine phosphatase receptor type O PTPase U2 PTP-oc PTP-U2glomerular epithelial protein-1 PTPU2osteoclastic transmembrane protein-tyrosine phosphatase receptor-type protein tyrosine phosphatase O receptor-type tyrosine-protein phosphatase O R-PTP-O; Glomerular epithelial protein 1; osteoclastic transmembrane protein-tyrosine phosphatase; phosphotyrosine phosphatase U2; protein tyrosine phosphatase PTP-U2; Protein tyrosine phosphatase U2; protein tyrosine phosphatase, receptor type, O; PTP phi; PTP-U2; PTPase U2; R-PTP-O; Receptor-type tyrosine-protein phosphatase O
Gene Aliases: GLEPP1; NPHS6; PTP-OC; PTP-U2; PTPRO; PTPROT; PTPU2; R-PTP-O
UniProt ID: (Human) Q16827
Entrez Gene ID: (Human) 5800
Molecular Function:
hydrolase
phosphatase
protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.