Proteintech
Manufacturer Code:121611AP150UL
Catalog # 50-172-7758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PTPRO Fusion Protein Ag2802 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GLEPP1 PTP U2 PTPase U2 PTPRO PTPU2 R PTP O; Glomerular epithelial protein 1; osteoclastic transmembrane protein-tyrosine phosphatase; phosphotyrosine phosphatase U2; protein tyrosine phosphatase PTP-U2; Protein tyrosine phosphatase U2; protein tyrosine phosphatase, non-receptor type 15; protein tyrosine phosphatase, receptor type, O; PTP phi; PTP-U2; PTPase U2; R-PTP-O; receptor protein tyrosine phosphatase RO; receptor-type protein tyrosine phosphatase D30; Receptor-type tyrosine-protein phosphatase O
Gene Aliases: D28; GLEPP1; NPHS6; PTP-OC; PTP-U2; PTPBK; Ptpn15; PTPoc; PTPphi; PTPRO; PTPROT; PTPU2; R-PTP-O; RPTP-BK
UniProt ID: (Human) Q16827, (Mouse) E9Q612
Entrez Gene ID: (Human) 5800, (Rat) 50677, (Mouse) 19277
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.