Invitrogen
Manufacturer Code:PA518906
Catalog # PIPA518906
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (QGEGNIKKNRHPD) corresponding to the internal amino acids of PTPRN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: IA-2 protein tyrosine phosphatase receptor type N FLJ16131 IA-2/PTP IA2 ICA512 R-PTP-N islet antigen 2 islet cell antigen 512 islet cell autoantigen 3 protein tyrosine phosphatase-like N; protein tyrosine phosphatase, receptor-type, N; PTP IA-2; R-PTP-N; receptor-type tyrosine-protein phosphatase-like N
Gene Aliases: IA-2; mIA-A; mKIAA4064
Entrez Gene ID: (Mouse) 19275
Molecular Function:
hydrolase
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.