Invitrogen
Manufacturer Code:MA531873
Catalog # PIMA531873
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5H5D7 |
Immunogen | Purified recombit fragment of human PTPRD (AA: extra 1077-1265) expressed in E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HPTP PTPD HPTPD HPTPDELTA RPTPDELTA; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, delta polypeptide; Protein-tyrosine phosphatase delta; R-PTP-delta; Receptor-type tyrosine-protein phosphatase delta
Gene Aliases: HPTP; HPTPD; HPTPDELTA; PTPD; PTPRD; RPTPDELTA
UniProt ID: (Human) P23468
Entrez Gene ID: (Human) 5789
Molecular Function:
hydrolase
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.