Novus Biologicals
Manufacturer Code:NBP29339620UL
Catalog # NB028675
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1265 of human PTPRD (NP_002830.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HPTPDELTA HPTPMGC119752 MGC119750 MGC119751 protein tyrosine phosphatase receptor type D Protein-tyrosine phosphatase delta PTPDMGC119753 receptor type delta polypeptide receptor-type tyrosine-protein phosphatase delta; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, delta polypeptide; Protein-tyrosine phosphatase delta; R-PTP-delta; Receptor-type tyrosine-protein phosphatase delta
Gene Aliases: HPTP; HPTPD; HPTPDELTA; PTPD; PTPRD; RPTPDELTA
UniProt ID: (Human) P23468
Entrez Gene ID: (Human) 5789
Molecular Function: hydrolase phosphatase protein phosphatase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.