Novus Biologicals
Manufacturer Code:NBP281772
Catalog # NB017083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PTPRD antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PTPRD. The immunogen is located within the first 50 amino acids of PTPRD. Amino Acid Squence: RTDAETPPRFTRTPVDQT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HPTPDELTA HPTPMGC119752 MGC119750 MGC119751 protein tyrosine phosphatase receptor type D Protein-tyrosine phosphatase delta PTPDMGC119753 receptor type delta polypeptide receptor-type tyrosine-protein phosphatase delta; protein tyrosine phosphatase, receptor type, D; protein tyrosine phosphatase, receptor type, delta polypeptide; Protein-tyrosine phosphatase delta; R-PTP-delta; Receptor-type tyrosine-protein phosphatase delta
Gene Aliases: HPTP; HPTPD; HPTPDELTA; PTPD; PTPRD; RPTPDELTA
UniProt ID: (Human) P23468
Entrez Gene ID: (Human) 5789
Molecular Function:
hydrolase
phosphatase
protein phosphatase
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.