Novus Biologicals
Manufacturer Code:NBP256421
Catalog # NBP256421
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:HPLPSYLEYRHNASIRVYQTNYFASKCAENPNSNSKSFNIKLGAEMESLGGKCDPTQQKFCDGPLKPHTAYRISIRAFTQLFDEDLKEFTKPLYSD |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686E2262 DKFZp686H15164 EC 3.1.3.48 FLJ44133 HPTPB HPTP-BETA MGC142023 MGC59935 protein tyrosine phosphatase receptor type B protein tyrosine phosphatase receptor type beta polypeptide Protein-tyrosine phosphatase beta PTPB receptor-type tyrosine-protein phosphatase beta R-PTP-BETA Vascular endothelial protein tyrosine phosphatase VEPTP VE-PTP; protein tyrosine phosphatase, receptor type, B; protein tyrosine phosphatase, receptor type, beta polypeptide; Protein-tyrosine phosphatase beta; Receptor-type tyrosine-protein phosphatase beta; Vascular endothelial protein tyrosine phosphatase; VE-PTP
Gene Aliases: HPTP-BETA; HPTPB; PTPB; PTPRB; R-PTP-BETA; VEPTP
UniProt ID: (Human) P23467
Entrez Gene ID: (Human) 5787
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.