Abnova
Manufacturer Code:MAB19076
Catalog # 89-335-910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 2A7 |
Immunogen | A synthetic phosphopeptide corresponding to residues surrounding Y536 of human PTPN6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: HCP; HCPH; hematopoietic cell phosphatase; Hematopoietic cell protein-tyrosine phosphatase; HPTP1C; Protein-tyrosine phosphatase 1C; Protein-tyrosine phosphatase SHP-1; PTP-1C; SH-PTP1; SHP-1; SHP-1L; SHP1; Tyrosine-protein phosphatase non-receptor type 6
Gene Aliases: HCP; HCPH; HPTP1C; PTP-1C; PTP1C; PTPN6; SH-PTP1; SHP-1; SHP-1L; SHP1
UniProt ID: (Human) P29350
Entrez Gene ID: (Human) 5777
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.