Proteintech
Manufacturer Code:104721AP150UL
Catalog # 50-172-6332
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PTPN23 Fusion Protein Ag0765 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HD PTP HDPTP KIAA1471 PTP TD14 PTPN23; HD-PTP; His domain-containing protein tyrosine phosphatase; Protein tyrosine phosphatase TD14; PTP-TD14; Tyrosine-protein phosphatase non-receptor type 23
Gene Aliases: AI462446; HD-PTP; HDPTP; KIAA1471; PTP-TD14; PTPN23
UniProt ID: (Human) Q9H3S7, (Mouse) Q6PB44
Entrez Gene ID: (Human) 25930, (Mouse) 104831, (Rat) 117552
Molecular Function:
hydrolase
phosphatase
protein phosphatase
receptor
transmembrane receptor regulatory/adaptor protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.