Proteintech
Manufacturer Code:117831AP150UL
Catalog # 50-172-7451
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PTPN22 Fusion Protein Ag2397 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hematopoietic cell protein-tyrosine phosphatase 70Z-PEP; Lymphoid phosphatase; Lymphoid phosphatase LYP LYP1 LYP2 PEP PTPN22 PTPN8; lymphoid-specific protein tyrosine phosphatase; LyP; PEP; PEST domain-enriched tyrosine phosphatase; PEST-domain phosphatase; protein tyrosine phosphatase, non-receptor type 22 (lymphoid); protein tyrosine phosphatase, non-receptor type 8; Tyrosine-protein phosphatase non-receptor type 22
Gene Aliases: 70zpep; LYP; LYP1; LYP2; PEP; PTPN22; PTPN8
UniProt ID: (Human) Q9Y2R2, (Mouse) P29352
Entrez Gene ID: (Human) 26191, (Rat) 295338, (Mouse) 19260
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.