Invitrogen
Manufacturer Code:MA517249
Catalog # PIMA517249
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | T25-AF3B7 |
Immunogen | Recombit human protein purified from E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: PTPT T-cell protein-tyrosine phosphatase Tyrosine-protein phosphatase non-receptor type 2 PTN2; T-cell protein tyrosine phosphatase; T-cell protein-tyrosine phosphatase; TCPTP; Tyrosine-protein phosphatase non-receptor type 2
Gene Aliases: PTN2; PTPN2; PTPT; TC-PTP; TCELLPTP; TCPTP
UniProt ID: (Human) P17706
Entrez Gene ID: (Human) 5771
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.