Novus Biologicals
Manufacturer Code:NBP261809
Catalog # NBP2-61809
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B3H7 |
Immunogen | Purified recombinant fragment of human PTPN14 (AA: 896-1169) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
Protein Aliases: cytoskeletal-associated protein tyrosine phosphatase; EC 3.1.3 EC 3.1.3.48 MGC126803 PEZcytoskeletal-associated protein tyrosine phosphatase protein tyrosine phosphatase non-receptor type 14 Protein-tyrosine phosphatase pez PTP36 tyrosine-protein phosphatase non-receptor type 14; Protein-tyrosine phosphatase pez; Tyrosine-protein phosphatase non-receptor type 14
Gene Aliases: PEZ; PTP36; PTPD2; PTPN14
UniProt ID: (Human) Q15678
Entrez Gene ID: (Human) 5784
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.