OriGene
Manufacturer Code:TA501759
Catalog # 50-167-2877
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F4 |
Immunogen | Full length human recombit protein of human PTPN11 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BPTP3 CFC JMML METCDS NS1 PTP-1D PTP2C SH-PTP2 SH-PTP3 SHP2; Protein-tyrosine phosphatase 1D; Protein-tyrosine phosphatase 2C; PTP-1D; PTP-2C; SH-PTP2; SH-PTP3; SHP-2; Tyrosine-protein phosphatase non-receptor type 11
Gene Aliases: BPTP3; CFC; JMML; METCDS; NS1; PTP-1D; PTP2C; PTPN11; SH-PTP2; SH-PTP3; SHP2; SHPTP2
UniProt ID: (Human) Q06124
Entrez Gene ID: (Human) 5781
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.