Abnova
Manufacturer Code:MAB8799
Catalog # 89-113-589
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 68r78 |
Immunogen | Recombinant protein corresponding to full length human PTPMT1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against full-length recombinant PTPMT1.
For Research Use Only
Protein Aliases: DUSP23; FLJ46081; MOSP; NB4 apoptosis/differentiation related protein; Phosphatidylglycerophosphatase and protein-tyrosine phosphatase 1; Phosphoinositide lipid phosphatase; PLIP; PNAS-129; Protein-tyrosine phosphatase mitochondrial 1; PTEN-like phosphatase
Gene Aliases: DUSP23; MOSP; PLIP; PNAS-129; PTPMT1
UniProt ID: (Human) Q8WUK0
Entrez Gene ID: (Human) 114971
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.