Invitrogen
Manufacturer Code:PA5113050
Catalog # PIPA5113050
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2 protein (7-41AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxyacyl-CoA dehydratase 2; 3-hydroxyacyl-CoA dehydratase 2 6330408J20Rik AI255777 AI481689 Hacd2 Hcad2 I79_000662 protein tyrosine phosphatase-like (proline instead of catalytic arginine) member b Protein-tyrosine phosphatase-like member B PTPLB very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 2 Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2; HACD2; protein tyrosine phosphatase-like (proline instead of catalytic arginine), member b; Protein-tyrosine phosphatase-like member B; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 2; Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2
Gene Aliases: HACD2; PTPLB
UniProt ID: (Human) Q6Y1H2
Entrez Gene ID: (Human) 201562
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.