Abnova
Manufacturer Code:H00201562K
Catalog # 89-208-612
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PTPLB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human PTPLB peptide using ARM Technology.
For Research Use Only
Protein Aliases: 3-hydroxyacyl-CoA dehydratase 2; HACD2; protein tyrosine phosphatase-like (proline instead of catalytic arginine), member b; Protein-tyrosine phosphatase-like member B; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 2; Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 2
Gene Aliases: HACD2; PTPLB
UniProt ID: (Human) Q6Y1H2
Entrez Gene ID: (Human) 201562
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.