R&D Systems
Manufacturer Code:MAB13662100
Catalog # MAB13662100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 208607 |
Immunogen | <em>E. coli</em>-derived recombinant human PTP1B Glu2-Thr435 Accession # P18031 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: EC 3.1.3.48 protein tyrosine phosphatase 1B protein tyrosine phosphatase non-receptor type 1 Protein-tyrosine phosphatase 1B PTP-1B PTP1Bprotein tyrosine phosphatase placental PTPN1 tyrosine-protein phosphatase non-receptor type 1; protein tyrosine phosphatase, placental; Protein-tyrosine phosphatase 1B; PTP-1B; Tyrosine-protein phosphatase non-receptor type 1
Gene Aliases: PTP1B; PTPN1
UniProt ID: (Human) P18031
Entrez Gene ID: (Human) 5770
Molecular Function: hydrolase phosphatase protein phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.