Invitrogen
Manufacturer Code:PA529312
Catalog # PIPA529312
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 92 and 338 of Human PTK9 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A6 protein tyrosine kinase; A6 PTK9 A6 protein tyrosine kinase PTK9 protein tyrosine kinase 9 protein A6 protein tyrosine kinase 9 twinfilin-1; actin monomer-binding protein; Protein A6; Protein tyrosine kinase 9; PTK9 protein tyrosine kinase 9; twinfilin actin-binding protein 1; twinfilin, actin-binding protein, homolog 1; Twinfilin-1
Gene Aliases: A6; PTK9; TWF1; twinfilin
UniProt ID: (Human) Q12792, (Mouse) Q91YR1
Entrez Gene ID: (Human) 5756, (Mouse) 19230
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.