Novus Biologicals
Manufacturer Code:NBP273708PECY7
Catalog # N273708PEC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2E7 |
Immunogen | Full length recombinant protein of human PTK7/CCK4 (NP_690619) produced in HEK293T cells. |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: CCK-4; CCK-4 CCK4Colon carcinoma kinase 4 colon carcinoma kinase-4 Protein-tyrosine kinase 7 Pseudo tyrosine kinase receptor 7 PTK7 protein tyrosine kinase 7 Tyrosine-protein kinase-like 7; Colon carcinoma kinase 4; Inactive tyrosine-protein kinase 7; Protein-tyrosine kinase 7; Pseudo tyrosine kinase receptor 7; PTK7 protein tyrosine kinase 7; Tyrosine-protein kinase-like 7
Gene Aliases: CCK-4; CCK4; PTK7
UniProt ID: (Human) Q13308
Entrez Gene ID: (Human) 5754
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
protein kinase receptor
receptor
transferase
tyrosine protein kinase receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.