Invitrogen
Manufacturer Code:PA540799
Catalog # PIPA540799
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human PTHLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: calciotropic hormone, growth factor; Osteostatin; parathyroid hormone-like hormone; Parathyroid hormone-like hormone HHM PLP BDE2 PTHR PTHRP; parathyroid hormone-like hormone preproprotein; parathyroid hormone-like peptide; Parathyroid hormone-like protein; parathyroid hormone-like related protein; parathyroid hormone-related peptide; Parathyroid hormone-related protein; parathyroid-like peptide; PLP; PTH-related peptide; PTH-related protein; PTH-rP; PTHrP; PTHrP[1-36]; PTHrP[107-139]; PTHrP[38-94]
Gene Aliases: BDE2; HHM; PLP; PTH-like; PTH-rP; PTHLH; PTHR; PTHRP
UniProt ID: (Human) Q6FH74, (Bovine) P58073, (Dog) P52211, (Rabbit) Q9GLC7, (Rat) P13085
Entrez Gene ID: (Goat) 100860907, (Human) 5744, (Bovine) 286767, (Horse) 100033979, (Dog) 403987, (Rabbit) 100009357, (Guinea pig) 100716151, (Rat) 24695, (Mouse) 19227
Molecular Function: peptide hormone signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.