Invitrogen
Manufacturer Code:PA577690
Catalog # PIPA577690
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide CFLQPDINIGKQE corresponding to amino acid residues 125-137 of rat Parathyroid Hormone 2 Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Parathyroid Hormone 2 Receptor; parathyroid hormone receptor 2; PTH2 receptor
Gene Aliases: Pth2r; Pthr2
UniProt ID: (Rat) P70555, (Mouse) Q91V95
Entrez Gene ID: (Rat) 81753, (Mouse) 213527
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.