Invitrogen
Manufacturer Code:PA534133
Catalog # PIPA534133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from 1st extracellular domain of human PTH2R / PTHR2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Parathyroid hormone 2 receptor; parathyroid hormone receptor 2; PTH2 receptor; PTH2R Parathyroid hormone 2 receptor Parathyroid hormone receptor 2 PTH2 receptor PTHR2 Hpth2 receptor
Gene Aliases: PTH2R; PTHR2
UniProt ID: (Human) P49190
Entrez Gene ID: (Human) 5746
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.