Novus Biologicals
Manufacturer Code:NBP294871100UL
Catalog # NB024250
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 27-145 of human PTH2R (NP_005039.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Parathyroid hormone 2 receptor; parathyroid hormone 2 receptor PTH2 receptor PTHR2parathyroid hormone receptor 2; parathyroid hormone receptor 2; PTH2 receptor
Gene Aliases: PTH2R; PTHR2
UniProt ID: (Human) P49190
Entrez Gene ID: (Human) 5746
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.