Invitrogen
Manufacturer Code:PA577689
Catalog # PIPA577689
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)DAVLYSGFTLDEAER corresponding to amino acid residues 241-255 of rat Parathyroid Hormone 1 Receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Parathyroid hormone 1 receptor; parathyroid hormone receptor 1; Parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PTH-related peptide receptor; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH/PTHrP type I receptor PTH1 receptor PTHR PTHR1; PTH1 receptor; seven transmembrane helix receptor
Gene Aliases: PFE; PPR; PTH1R; PTHR; PTHR1; PTHrel
UniProt ID: (Human) Q03431, (Rat) P25961, (Mouse) P41593
Entrez Gene ID: (Human) 5745, (Rat) 56813, (Mouse) 19228
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.