R&D Systems
Manufacturer Code:MAB5709SP
Catalog # MAB5709SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 734630 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human PTH1R/PTHR1 Tyr23-Met189 Accession # Q03431 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC138426 parathyroid hormone 1 receptorMGC138452 parathyroid hormone/parathyroid hormone-related peptide receptor parathyroid hormone/parathyroid hormone-related protein receptor PFE PTH/PTHr receptor PTH/PTHrP type I receptor PTH1 receptor PTHR PTHR1 PTHRPTHR1parathyroid hormone receptor 1 seven transmembrane helix receptor; Parathyroid hormone 1 receptor; parathyroid hormone receptor 1; Parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PTH/PTHr receptor; PTH/PTHrP type I receptor; PTH1 receptor; seven transmembrane helix receptor
Gene Aliases: PFE; PTH1R; PTHR; PTHR1
UniProt ID: (Human) Q03431
Entrez Gene ID: (Human) 5745
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.