Novus Biologicals
Manufacturer Code:NBP290079AF594
Catalog # NB069622
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 3 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human PTH1R/PTHR1 (Q03431 Met1-Gly188). |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC138426 parathyroid hormone 1 receptorMGC138452 parathyroid hormone/parathyroid hormone-related peptide receptor parathyroid hormone/parathyroid hormone-related protein receptor PFE PTH/PTHr receptor PTH/PTHrP type I receptor PTH1 receptor PTHRPTHR1parathyroid hormone receptor 1 seven transmembrane helix receptor; Parathyroid hormone 1 receptor; parathyroid hormone receptor 1; Parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PTH/PTHr receptor; PTH/PTHrP type I receptor; PTH1 receptor; seven transmembrane helix receptor
Gene Aliases: PFE; PTH1R; PTHR; PTHR1
UniProt ID: (Human) Q03431
Entrez Gene ID: (Human) 5745
Molecular Function: G-protein coupled receptor antibacterial response protein defense/immunity protein receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.