Abnova
Manufacturer Code:MAB10307
Catalog # 89-108-494
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D2 |
Immunogen | Recombinant protein corresponding to human PTH1R. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against recombinant PTH1R.
For Research Use Only
Protein Aliases: MGC138426; MGC138452; Parathyroid hormone 1 receptor; parathyroid hormone receptor 1; Parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PTH/PTHr receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTHR; PTHR1; seven transmembrane helix receptor
Gene Aliases: PFE; PTH1R; PTHR; PTHR1
UniProt ID: (Human) Q03431
Entrez Gene ID: (Human) 5745
Molecular Function:
G-protein coupled receptor
antibacterial response protein
defense/immunity protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.