Invitrogen
Manufacturer Code:PA577695
Catalog # PIPA577695
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)TQAIAPDSREMGD corresponding to amino acid residues 290-302 of rat Prostanoid Prostacyclin receptor |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PGI receptor; PGI2 receptor; Prostacyclin receptor; prostaglandin I receptor (IP); Prostaglandin I2 receptor; Prostanoid IP receptor; Prostanoid Prostacyclin Receptor (extracellular)
Gene Aliases: IP; PGI2; PRIPR; PTGIR
UniProt ID: (Human) P43119, (Mouse) P43252, (Rat) P43253
Entrez Gene ID: (Human) 5739, (Mouse) 19222, (Rat) 292661
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.