Invitrogen
Manufacturer Code:PA572393
Catalog # PIPA572393
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | region 221-245 aa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PGI receptor; PGI2 receptor; Prostacyclin receptor; Prostaglandin I2 receptor; Prostanoid IP receptor; PTGIR IP receptor PGI receptor PRIPR Prostaglandin I2 receptor IP PGI2 receptor Prostacyclin IP receptor Prostacyclin receptor Prostanoid IP receptor
Gene Aliases: IP; PRIPR; PTGIR
UniProt ID: (Human) P43119
Entrez Gene ID: (Human) 5739
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.