Invitrogen
Manufacturer Code:PA5102240
Catalog # PIPA5102240
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Prostacyclin Receptor corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IP IP receptor PGI receptor PGI2 PGI2 receptor PRIPR Prostacyclin receptor prostaglandin I receptor (IP) prostaglandin I2 (prostacyclin) receptor (IP) prostaglandin I2 receptor Prostanoid IP receptor Ptgir; PGI receptor; PGI2 receptor; Prostacyclin receptor; Prostaglandin I2 receptor; Prostanoid IP receptor
Gene Aliases: IP; PRIPR; PTGIR
UniProt ID: (Human) P43119
Entrez Gene ID: (Human) 5739
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.