Abnova
Manufacturer Code:PAB25121
Catalog # 89-126-795
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to PTGIR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PTGIR.
For Research Use Only
Protein Aliases: IP; MGC102830; PGI receptor; PGI2 receptor; PRIPR; Prostacyclin receptor; Prostaglandin I2 receptor; Prostanoid IP receptor
Gene Aliases: IP; PRIPR; PTGIR
UniProt ID: (Human) P43119
Entrez Gene ID: (Human) 5739
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.