Invitrogen
Manufacturer Code:PA570674
Catalog # PIPA570674
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C-terminal region of human PTGFR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FP prostanoid receptor; FP receptor; PGF receptor; PGF2 alpha receptor; PGF2-alpha receptor; prostaglandin F receptor (FP); prostaglandin F2 alpha receptor; Prostaglandin F2-alpha receptor; prostaglandin F2alpha receptor; prostaglandin receptor (2-alpha); Prostanoid FP receptor; PTGFR PGF receptor PGF2-alpha receptor Prostaglandin F receptor (FP) FP PGF2 alpha receptor Prostaglandin F receptor Prostanoid FP receptor
Gene Aliases: AI957154; FP; PF2AR; PGF; PTGFR
UniProt ID: (Bovine) P37289, (Sheep) Q28905, (Human) P43088, (Rat) P43118, (Mouse) P43117
Entrez Gene ID: (Bovine) 282020, (Sheep) 443366, (Dog) 479981, (Pig) 397126, (Horse) 100009714, (Human) 5737, (Rabbit) 100344670, (Rat) 25652, (Mouse) 19220, (Guinea pig) 100735880
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.