Invitrogen
Manufacturer Code:PA534128
Catalog # PIPA534128
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 16 amino acid peptide from 2nd extracellular domain of human PTGFR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PGF receptor; PGF2 alpha receptor; PGF2-alpha receptor; prostaglandin F receptor (FP); prostaglandin F2 alpha receptor; Prostaglandin F2-alpha receptor; prostaglandin receptor (2-alpha); Prostanoid FP receptor; PTGFR PGF receptor PGF2-alpha receptor Prostaglandin F receptor (FP) FP PGF2 alpha receptor Prostaglandin F receptor Prostanoid FP receptor
Gene Aliases: FP; PTGFR
UniProt ID: (Human) P43088
Entrez Gene ID: (Human) 5737
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.