Novus Biologicals
Manufacturer Code:NBP2823180.1ML
Catalog # NB030247
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human PTGFR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FP MGC120498 MGC46203 PGF receptor PGF2 alpha receptor PGF2-alpha receptor prostaglandin F receptor (FP) prostaglandin F2 alpha receptor prostaglandin F2-alpha receptor prostaglandin receptor (2-alpha) Prostanoid FP receptor; PGF receptor; PGF2 alpha receptor; PGF2-alpha receptor; prostaglandin F receptor (FP); prostaglandin F2 alpha receptor; Prostaglandin F2-alpha receptor; prostaglandin receptor (2-alpha); Prostanoid FP receptor
Gene Aliases: FP; PTGFR
UniProt ID: (Human) P43088
Entrez Gene ID: (Human) 5737
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.