Abnova
Manufacturer Code:PAB26512
Catalog # 89-128-152
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 18 amino acids at 1st extracellular domain of human PTGFR. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PTGFR.
For Research Use Only
Protein Aliases: FP; MGC120498; MGC46203; PGF receptor; PGF2 alpha receptor; PGF2-alpha receptor; prostaglandin F receptor (FP); prostaglandin F2 alpha receptor; Prostaglandin F2-alpha receptor; prostaglandin receptor (2-alpha); Prostanoid FP receptor
Gene Aliases: FP; PTGFR
UniProt ID: (Human) P43088
Entrez Gene ID: (Human) 5737
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.