Invitrogen
Manufacturer Code:PA587300
Catalog # PIPA587300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human PTGES2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: gamma-interferon-activated transcriptional element-binding factor 1; GATE-binding factor 1; GBF-1; membrane-associated prostaglandin E synthase 2; Membrane-associated prostaglandin E synthase-2; Microsomal prostaglandin E synthase 2; microsomal prostaglandin E synthase-2; mPGE synthase-2; mPGES-2; Prostaglandin E synthase 2; Prostaglandin E synthase 2 Membrane-associated prostaglandin E synthase-21 Publication mPGE synthase-21 Publication Microsomal prostaglandin E synthase 2 mPGES-2 Prostaglandin-H(2) E-isomerase PTGES2 C9orf15 PGES2; Prostaglandin E synthase 2 truncated form; Prostaglandin-H(2) E-isomerase
Gene Aliases: 0610038H10Rik; C79137; C9orf15; GBF-1; GBF1; mPGES-2; Mpges2; PGES2; PTGES2
UniProt ID: (Human) Q9H7Z7, (Mouse) Q8BWM0
Entrez Gene ID: (Human) 80142, (Rat) 311865, (Mouse) 96979
Molecular Function: oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.