Invitrogen
Manufacturer Code:PA535259
Catalog # PIPA535259
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 242-269 amino acids from the central region of human PTGER4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PGE receptor EP4 subtype; PGE receptor, EP4 subtype; PGE2 receptor EP4 subtype; prostaglandin E receptor 4 (subtype EP4); Prostaglandin E2 receptor EP4 subtype; Prostanoid EP4 receptor; PTGER4PTGER2 Prostaglandin E2 receptor EP4 subtype Prostaglandin E2 receptor EP4 subtype Prostanoid EP4 receptor
Gene Aliases: EP4; EP4R; PTGER2; PTGER4; Ptgerep4
UniProt ID: (Human) P35408, (Mouse) P32240
Entrez Gene ID: (Human) 5734, (Mouse) 19219
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.