Invitrogen
Manufacturer Code:PA5102057
Catalog # PIPA5102057
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PE2R3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EP3 Ep3 receptor EP3A receptor EP3B receptor EP3C receptor EP3D receptor EP3e EP3-I EP3-II EP3-III EP3-IV EP3-VI MGC141828 MGC141829 MGC27302 PGE receptor EP3 subtype PGE receptor EP3 subtype PGE2 receptor EP3 subtype PGE2-R Pgerep3 pr prostaglandin E receotor EP3 subtype 3 isoform prostaglandin E receptor 3 prostaglandin E receptor 3 (subtype EP3) prostaglandin E receptor EP3 subtype prostaglandin E2 receptor EP3 subtype prostaglandin receptor (PGE-2) prostanoid EP3 receptor Ptger3 Ptgerep3 Rep3 rEP3a rEP3b; PGE receptor EP3 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP3 subtype; PGE2-R; prostaglandin E receotor EP3 subtype 3 isoform; prostaglandin E receptor 3 (subtype EP3); Prostaglandin E2 receptor EP3 subtype; prostaglandin receptor (PGE-2); Prostanoid EP3 receptor
Gene Aliases: EP3; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; EP3e; PGE2-R; PTGER3
UniProt ID: (Human) P43115
Entrez Gene ID: (Human) 5733
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.