Abnova
Manufacturer Code:PAB26522
Catalog # 89-128-162
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids at 1st cytoplasmic domain of human PTGER3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of PTGER3.
For Research Use Only
Protein Aliases: EP3; EP3-I; EP3-II; EP3-III; EP3-IV; EP3e; MGC141828; MGC141829; MGC27302; PGE receptor EP3 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP3 subtype; PGE2-R; prostaglandin E receotor EP3 subtype 3 isoform; prostaglandin E receptor 3 (subtype EP3); Prostaglandin E2 receptor EP3 subtype; prostaglandin receptor (PGE-2); Prostanoid EP3 receptor
Gene Aliases: EP3; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; EP3e; PGE2-R; PTGER3
UniProt ID: (Human) P43115
Entrez Gene ID: (Human) 5733
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.