Invitrogen
Manufacturer Code:MA112677
Catalog # 01-674-033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F12 |
Immunogen | Recombit human EP2 and EP3 receptor protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EP2 receptor; PGE receptor EP2 subtype; PGE receptor EP3 subtype; PGE receptor, EP2 subtype; PGE receptor, EP3 subtype; PGE2 receptor EP2 subtype; PGE2 receptor EP3 subtype; PGE2-R; prostaglandin E receotor EP3 subtype 3 isoform; prostaglandin E receptor 2 (EP2 subtype); prostaglandin E receptor 2 (subtype EP2), 53kD; prostaglandin E receptor 2 (subtype EP2), 53kDa; prostaglandin E receptor 3 (subtype EP3); prostaglandin E receptor EP2 subtype; prostaglandin E receptor EP3 subtype; prostaglandin E receptor subtype EP2; Prostaglandin E2 receptor EP2 subtype; Prostaglandin E2 receptor EP2 subtype PGE receptor EP2 subtype PGE2 receptor EP2 subtype Prostanoid EP2 receptor PTGER2 Prostaglandin E2 receptor EP3 subtype PGE receptor EP3 subtype PGE2 receptor EP3 subtype PGE2-R Prostanoid EP3 receptor PTGER3 Prostaglandin E2 Receptor EP2 + EP3; Prostaglandin E2 receptor EP3 subtype; prostaglandin E2 receptor type 2; prostaglandin receptor (PGE-2); Prostanoid EP2 receptor; Prostanoid EP3 receptor
Gene Aliases: EP2; EP3; EP3-I; EP3-II; EP3-III; EP3-IV; EP3-VI; EP3e; PGE2-R; Pgerep3; Ptger-ep2; PTGER2; PTGER3; Ptgerep2; Ptgerep3; Rep3; rEP3a; rEP3b
UniProt ID: (Human) P43116, (Human) P43115, (Rat) Q62928, (Mouse) Q62053, (Rat) P34980
Entrez Gene ID: (Human) 5732, (Human) 5733, (Rat) 81752, (Mouse) 19217, (Rat) 24929, (Mouse) 19218
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.