Invitrogen
Manufacturer Code:PA591872
Catalog # PIPA591872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human PTGER2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EP2 receptor; PGE receptor EP2 subtype; PGE receptor, EP2 subtype; PGE2 receptor EP2 subtype; prostaglandin E receptor 2 (EP2 subtype); prostaglandin E receptor 2 (subtype EP2), 53kD; prostaglandin E receptor 2 (subtype EP2), 53kDa; prostaglandin E receptor EP2 subtype; prostaglandin E receptor subtype EP2; Prostaglandin E2 receptor EP2 subtype; prostaglandin E2 receptor type 2; Prostanoid EP2 receptor
Gene Aliases: EP2; Ptger-ep2; PTGER2; Ptgerep2
UniProt ID: (Human) P43116, (Rat) Q62928, (Mouse) Q62053
Entrez Gene ID: (Human) 5732, (Rat) 81752, (Mouse) 19217
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.