Invitrogen
Manufacturer Code:PA534114
Catalog # PIPA534114
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 20 amino acid peptide from N-terminal extracellular domain of human Prostaglandin D2 Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PGD receptor; PGD2 receptor; Prostaglandin D2 receptor; prostaglandin D2 receptor (DP); Prostanoid DP receptor; PTGDR DP Dp receptor PGD receptor Prostaglandin D2 receptor (DP) Prostanoid DP receptor PTGDR1 PGD2 receptor Prostaglandin DP receptor AS1 ASRT1 Dp prostanoid receptor DP1 Prostaglandin D2 receptor
Gene Aliases: AS1; ASRT1; DP; DP1; PTGDR; PTGDR1
UniProt ID: (Human) Q13258
Entrez Gene ID: (Human) 5729
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.